siRNA Details
Gene Symbol: IQCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095094.1 | - | 2 | 1841 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK096574.1 | - | 1 | 67 | Chr. 3 - 197955287 - 197958085 [-] on Build GRCh38 |
