siRNA Details
Gene Symbol: LOC101929679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL357200.1 | - | 1 | 57 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | T81058.1 | - | 58 | Chr. 6 - 135181370 - 135181386 [+] on Build GRCh38 |
