siRNA Details
Gene Symbol: LIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA872458.1 | - | 1 | 362 | ||
AW166526.1 | - | 1 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK127668.1 | - | 1 | 936 | Chr. 17 - 35003318 - 35005642 [-] on Build GRCh38 |
