siRNA Details
Gene Symbol: LINC01361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110632.1 | 3 | 530 | |||
NR_110633.1 | 3 | 527 | ||||
GenBank mRNA | BC037222.1 | - | 3 | 530 | ||
U80773.1 | - | 3 | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI488658.1 | - | 3 | 530 | Chr. 1 - 82974506 - 82984922 [-] on Build GRCh38 |
