siRNA Details
Gene Symbol: LOC100507437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125384.1 | 3 | 1385 | |||
NR_125385.1 | 2 | 1250 | ||||
GenBank mRNA | AK123017.1 | - | 2 | 1250 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125384.1 | 3 | 1385 | |||
NR_125385.1 | 2 | 1250 | ||||
GenBank mRNA | AK123017.1 | - | 2 | 1250 |