siRNA Details
Gene Symbol: RBM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091279.1 | - | 1 | 2146 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125665.1 | - | 2 | 2843 | Chr. 1 - 110368399 - 110373003 [+] on Build GRCh38 |
