siRNA Details
Gene Symbol: HIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358103.1 | - | 1 | 53 | ||
JX965370.1 | - | 2 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR745472.1 | - | 2 | 221 | Chr. 7 - 75733460 - 75738962 [-] on Build GRCh38 |
