siRNA Details
Gene Symbol: LINC01094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038303.1 | 5 | 2600 | |||
NR_038304.1 | 4 | 2528 | ||||
NR_038305.1 | 4 | 2506 | ||||
NR_038306.1 | 3 | 2434 | ||||
NR_038307.1 | 3 | 2419 | ||||
NR_038308.1 | 3 | 2380 | ||||
GenBank mRNA | BC040182.2 | - | 3 | 2153 | ||
BC044613.1 | - | 3 | 2432 |
