siRNA Details
Gene Symbol: LINC01094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038308.1 | 2, 3 | 317 | |||
GenBank mRNA | BC040182.2 | - | 2, 3 | 94 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM172453.1 | - | 2, 3 | 99 | Chr. 4 - 78646212 - 78681354 [+] on Build GRCh38 |
