siRNA Details
Gene Symbol: ISG20L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303095.1 | NP_001290024.1 | 4 | 2522 | ||
XM_005245516.4 | XP_005245573.1 | 4 | 2490 | |||
XM_005245518.4 | XP_005245575.1 | 3 | 2037 | |||
XM_006711560.3 | XP_006711623.1 | 4 | 2660 | |||
XM_006711561.3 | XP_006711624.1 | 4 | 2549 | |||
GenBank mRNA | CV573712.1 | - | 1 | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK127588.1 | - | 2536 | Chr. 5 - 72152757 - 72155246 [+] on Build GRCh38 |
