siRNA Details
Gene Symbol: LOC101928012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110158.1 | 2 | 275 | |||
GenBank mRNA | BC041883.1 | - | 2 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI561763.1 | - | 2 | 283 | Chr. 7 - 112622378 - 112699489 [+] on Build GRCh38 |
