siRNA Details
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 3035 | ||
NM_001322924.1 | NP_001309853.1 | 5 | 2963 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR597367.1 | - | 1 | 667 | Chr. 1 - 167705673 - 167707280 [+] on Build GRCh38 |
