siRNA Details
Gene Symbol: LINC01310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038944.1 | 4 | 361 | |||
GenBank mRNA | AK056844.1 | - | 4 | 361 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA865773.1 | - | 4 | 361 | Chr. 22 - 48866770 - 48892877 [+] on Build GRCh38 |
