siRNA Details
Gene Symbol: LINC01310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038944.1 | 6 | 1057 | |||
GenBank mRNA | AK056844.1 | - | 6 | 1057 | ||
AK124622.1 | - | 2 | 1712 | |||
BC037788.1 | - | 3 | 473 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038944.1 | 6 | 1057 | |||
GenBank mRNA | AK056844.1 | - | 6 | 1057 | ||
AK124622.1 | - | 2 | 1712 | |||
BC037788.1 | - | 3 | 473 |