siRNA Details
Gene Symbol: SIX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL540911.3 | - | 1 | 574 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR622406.1 | - | 1 | 574 | Chr. 19 - 45768387 - 45769685 [+] on Build GRCh38 |
