siRNA Details
Gene Symbol: LINC00951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038887.1 | 1 | 1028 | |||
GenBank mRNA | AK123643.1 | - | 1 | 1028 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126514.1 | - | 1 | 2172 | Chr. 2 - 99284599 - 99288398 [+] on Build GRCh38 |
