siRNA Details
Gene Symbol: LOC101927078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130785.1 | 13 | 1798 | |||
GenBank mRNA | AK097686.1 | - | 13 | 1793 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB515172.1 | - | 4 | 308 | Chr. 5 - 114447519 - 114472351 [-] on Build GRCh38 |
