siRNA Details
Gene Symbol: CCDC148-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038850.1 | 4 | 304 | |||
GenBank mRNA | BC021739.1 | - | 4 | 309 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG720090.1 | - | 4 | 309 | Chr. 2 - 158166650 - 158235978 [+] on Build GRCh38 |
