siRNA Details
Gene Symbol: DLGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003809.2 | NP_001003809.1 | 10 | 4506 | ||
NM_001242762.1 | NP_001229691.1 | 10 | 4449 | |||
NM_001242763.1 | NP_001229692.1 | 11 | 4563 | |||
NM_001242764.1 | NP_001229693.1 | 10 | 4479 | |||
NM_001242766.1 | NP_001229695.1 | 11 | 4536 | |||
NM_001308390.1 | NP_001295319.1 | 11 | 4561 | |||
NM_004746.3 | NP_004737.2 | 13 | 5863 | |||
XM_005258171.2 | XP_005258228.1 | 14 | 5934 | |||
XM_005258172.1 | XP_005258229.1 | 14 | 5929 | |||
XM_005258173.3 | XP_005258230.2 | 12 | 4725 | |||
XM_005258174.4 | XP_005258231.1 | 11 | 4782 | |||
XM_006722367.3 | XP_006722430.1 | 12 | 4893 | |||
XM_017026080.1 | XP_016881569.1 | 11 | 4862 | |||
XM_017026081.1 | XP_016881570.1 | 10 | 4526 | |||
XM_017026083.1 | XP_016881572.1 | 10 | 4751 | |||
GenBank mRNA | AK095761.1 | - | 1 | 2069 |
