siRNA Details
Gene Symbol: DLGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003809.2 | NP_001003809.1 | 10 | 3507 | ||
NM_001242762.1 | NP_001229691.1 | 10 | 3450 | |||
NM_001242763.1 | NP_001229692.1 | 11 | 3564 | |||
NM_001242764.1 | NP_001229693.1 | 10 | 3480 | |||
NM_001242766.1 | NP_001229695.1 | 11 | 3537 | |||
NM_001308390.1 | NP_001295319.1 | 11 | 3562 | |||
NM_004746.3 | NP_004737.2 | 13 | 4864 | |||
XM_005258171.2 | XP_005258228.1 | 14 | 4935 | |||
XM_005258172.1 | XP_005258229.1 | 14 | 4930 | |||
XM_005258173.3 | XP_005258230.2 | 12 | 3726 | |||
XM_005258174.4 | XP_005258231.1 | 11 | 3783 | |||
XM_006722367.3 | XP_006722430.1 | 12 | 3894 | |||
XM_017026080.1 | XP_016881569.1 | 11 | 3863 | |||
XM_017026081.1 | XP_016881570.1 | 10 | 3527 | |||
XM_017026083.1 | XP_016881572.1 | 10 | 3752 | |||
GenBank mRNA | AK095761.1 | - | 1 | 1070 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR001101.1 | - | 1 | 278 | Chr. 18 - 3497331 - 3498058 [-] on Build GRCh38 |
