siRNA Details
Gene Symbol: ZNF641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268637.3 | XP_005268694.1 | 5 | 5834 | ||
XM_005268638.3 | XP_005268695.1 | 6 | 6074 | |||
XM_005268639.2 | XP_005268696.1 | 5 | 5725 | |||
XM_011537897.1 | XP_011536199.1 | 6 | 6054 | |||
XM_011537898.1 | XP_011536200.1 | 6 | 5982 | |||
XM_017018797.1 | XP_016874286.1 | 5 | 5878 | |||
XM_017018798.1 | XP_016874287.1 | 6 | 5908 | |||
GenBank mRNA | AK054984.1 | - | 1 | 739 | ||
AK127710.1 | - | 1 | 1686 |
