siRNA Details
Gene Symbol: LOC101928535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120551.1 | 4, 5 | 350 | |||
GenBank mRNA | BC043242.1 | - | 4, 5 | 356 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI560262.1 | - | 4, 5 | 356 | Chr. 11 - 106251407 - 106263878 [+] on Build GRCh38 |
