siRNA Details
Gene Symbol: FLJ46875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128708.1 | - | 2 | 2747 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX096284.1 | - | 1 | 435 | Chr. 2 - 156338972 - 156339729 [-] on Build GRCh38 |
