siRNA Details
Gene Symbol: LINC00693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038840.1 | 5 | 1116 | |||
GenBank mRNA | BX369823.2 | - | 1 | 144 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR624823.1 | - | 1 | 345 | Chr. 3 - 28757455 - 28758810 [+] on Build GRCh38 |
