siRNA Details
Gene Symbol: LOC286382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094356.1 | - | 1 | 1678 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX095918.1 | - | 1 | 117 | Chr. 9 - 71059052 - 71059429 [-] on Build GRCh38 |
