siRNA Details
Gene Symbol: LINC01249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034134.1 | 4 | 407 | |||
GenBank mRNA | AK307324.1 | - | 4 | 407 | ||
BC047589.1 | - | 4 | 401 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034134.1 | 4 | 407 | |||
GenBank mRNA | AK307324.1 | - | 4 | 407 | ||
BC047589.1 | - | 4 | 401 |