siRNA Details
Gene Symbol: LOC101928936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110867.1 | 2 | 296 | |||
GenBank mRNA | AK131305.1 | - | 2 | 296 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA721939.1 | - | 2 | 296 | Chr. 6 - 88529337 - 88539368 [-] on Build GRCh38 |
