siRNA Details
Gene Symbol: ARMC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511910.1 | XP_011510212.1 | 25 | 3800 | ||
XM_011511912.1 | XP_011510214.1 | 24 | 3684 | |||
XM_017005022.1 | XP_016860511.1 | 25 | 3839 | |||
GenBank mRNA | BC014149.2 | - | 1 | 1056 |
