siRNA Details
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 494 | |||
GenBank mRNA | AK096958.1 | - | 1 | 460 | ||
BC104941.1 | - | 1 | 519 | |||
BC104943.1 | - | 1 | 519 | |||
DA532447.1 | - | 1 | 497 |
