siRNA Details
Gene Symbol: LOC101928877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY494058.1 | - | 5 | 963 | ||
BC043280.2 | - | 5 | 1111 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY494058.1 | - | 5 | 963 | ||
BC043280.2 | - | 5 | 1111 |