siRNA Details
Gene Symbol: LOC101927793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135194.1 | 1 | 41 | |||
GenBank mRNA | AK055866.1 | - | 1 | 284 | ||
DA400871.1 | - | 2 | 397 | |||
DA641322.1 | - | 2 | 168 | |||
DB173038.1 | - | 2 | 210 |
