siRNA Details
Gene Symbol: LOC101928877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY494057.1 | - | 3 | 259 | ||
BC043280.2 | - | 2 | 175 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI561957.1 | - | 2 | 180 | Chr. 4 - 185051896 - 185062797 [+] on Build GRCh38 |
