siRNA Details
Gene Symbol: LOC101928441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120471.1 | 3 | 600 | |||
GenBank mRNA | AK058180.1 | - | 3 | 588 | ||
AL517295.3 | - | 3 | 483 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI825627.1 | - | 3 | 545 | Chr. 12 - 23175642 - 23188519 [+] on Build GRCh38 |
