siRNA Details
Gene Symbol: SEC24D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125962.1 | - | 14 | 1993 | ||
BC037736.1 | - | 23 | 3479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK000883.1 | - | 1 | 1389 | Chr. 4 - 118736045 - 118737749 [-] on Build GRCh38 |
