siRNA Details
Gene Symbol: ZNF736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170905.2 | NP_001164376.1 | 4 | 6193 | ||
GenBank mRNA | AK126370.1 | - | 1 | 792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125478.1 | - | 1 | 2548 | Chr. 7 - 64351235 - 64354685 [+] on Build GRCh38 |
