siRNA Details
Gene Symbol: MIR205HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091113.1 | - | 1 | 2875 | ||
BC063704.1 | - | 1 | 102 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG697929.1 | - | 1 | 102 | Chr. 1 - 209431592 - 209432463 [+] on Build GRCh38 |
