siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 1 | 188 | ||
NM_001278663.1 | NP_001265592.1 | 1 | 188 | |||
NM_001278677.1 | NP_001265606.1 | 1 | 188 | |||
XM_011528443.1 | XP_011526745.1 | 1 | 177 | |||
XM_011528444.1 | XP_011526746.1 | 1 | 119 | |||
XM_011528448.2 | XP_011526750.1 | 1 | 79 | |||
XM_017027514.1 | XP_016883003.1 | 1 | 112 | |||
XM_017027515.1 | XP_016883004.1 | 1 | 176 | |||
XM_017027518.1 | XP_016883007.1 | 1 | 177 | |||
XM_017027519.1 | XP_016883008.1 | 1 | 176 | |||
GenBank mRNA | AK092080.1 | - | 1 | 144 | ||
AK092150.1 | - | 1 | 146 | |||
AK303431.1 | - | 1 | 122 | |||
BX281205.1 | - | 1 | 56 | |||
DA101675.1 | - | 1 | 106 |
