siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528443.1 | XP_011526745.1 | 2, 3 | 310 | ||
XM_011528448.2 | XP_011526750.1 | 2, 3 | 212 | |||
GenBank mRNA | AK092150.1 | - | 2, 3 | 279 |
