siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092080.1 | - | 7 | 892 | ||
AK092150.1 | - | 4 | 567 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA690246.1 | - | 4 | 568 | Chr. 19 - 24033445 - 24065626 [+] on Build GRCh38 |
