siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278677.1 | NP_001265606.1 | 3 | 369 | ||
XM_011528443.1 | XP_011526745.1 | 3 | 430 | |||
XM_011528444.1 | XP_011526746.1 | 4 | 450 | |||
XM_011528448.2 | XP_011526750.1 | 3 | 332 | |||
XM_017027514.1 | XP_016883003.1 | 3 | 360 | |||
XM_017027515.1 | XP_016883004.1 | 5 | 787 | |||
GenBank mRNA | AK092080.1 | - | 4 | 475 | ||
AK092150.1 | - | 3 | 399 | |||
DA101675.1 | - | 3 | 287 |
