siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528443.1 | XP_011526745.1 | 3 | 348 | ||
XM_011528444.1 | XP_011526746.1 | 4 | 368 | |||
XM_011528448.2 | XP_011526750.1 | 3 | 250 | |||
XM_017027514.1 | XP_016883003.1 | 3 | 278 | |||
XM_017027515.1 | XP_016883004.1 | 5 | 705 | |||
GenBank mRNA | AK092080.1 | - | 4 | 393 | ||
AK092150.1 | - | 3 | 317 |
