siRNA Details
Gene Symbol: LOC101928985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135066.1 | 2 | 2238 | |||
GenBank mRNA | BC057386.1 | - | 1 | 2581 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX101634.1 | - | 1 | 261 | Chr. 11 - 115731942 - 115732422 [-] on Build GRCh38 |
