siRNA Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274751.4 | XP_005274808.1 | 12 | 3499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK098449.1 | - | 1 | 316 | Chr. X - 155123316 - 155124935 [+] on Build GRCh38 |
