siRNA Details
Gene Symbol: EHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206615.1 | NP_001193544.1 | 8 | 5019 | ||
NM_001206616.1 | NP_001193545.1 | 9 | 5089 | |||
NM_012153.5 | NP_036285.2 | 9 | 5088 | |||
XM_005252857.1 | XP_005252914.1 | 9 | 5086 | |||
XM_005252860.1 | XP_005252917.1 | 9 | 4942 | |||
XM_005252861.1 | XP_005252918.1 | 8 | 5020 | |||
XM_005252862.1 | XP_005252919.1 | 8 | 4889 | |||
XM_011519983.1 | XP_011518285.1 | 9 | 5114 | |||
XM_011519984.1 | XP_011518286.1 | 10 | 5194 | |||
XM_011519985.1 | XP_011518287.1 | 9 | 4979 | |||
XM_017017534.1 | XP_016873023.1 | 8 | 5017 |
