siRNA Details
Gene Symbol: LOC101928591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135065.1 | 2 | 113 | |||
GenBank mRNA | AY726565.1 | - | 2 | 77 | ||
BX109663.1 | - | 2 | 113 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI756420.1 | - | 5 | 345 | Chr. 11 - 42068575 - 42891238 [-] on Build GRCh38 |
