siRNA Details
Gene Symbol: LOC100128198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125043.1 | - | 1 | 3162 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB331054.1 | - | 1 | 40 | Chr. 21 - 32280476 - 32280998 [-] on Build GRCh38 |
