siRNA Details
Gene Symbol: LOC101929269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134308.1 | 2 | 279 | |||
GenBank mRNA | BC038544.1 | - | 2 | 279 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI834352.1 | - | 279 | Chr. - - [-] on Build |
