siRNA Details
Gene Symbol: LOC105370888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135678.1 | 1 | 65 | |||
GenBank mRNA | BI520439.1 | - | 1 | 35 | ||
HY009381.1 | - | 1 | 66 | |||
HY016897.1 | - | 1 | 65 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK026340.1 | - | 1 | 994 | Chr. 15 - 72473836 - 72475168 [-] on Build GRCh38 |
