siRNA Details
Gene Symbol: ZNF316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278559.1 | NP_001265488.1 | 9 | 4687 | ||
XM_006715630.3 | XP_006715693.1 | 8 | 4804 | |||
GenBank mRNA | AL832182.1 | - | 1 | 906 |
