siRNA Details
Gene Symbol: LOC101927560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097722.1 | - | 2, 3 | 253 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB048258.1 | - | 2, 3 | 253 | Chr. 1 - 83802948 - 83860546 [-] on Build GRCh38 |
